


(±)-2-Methyltetrahydrofuran 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:96-47-9
:
Formula:C₅H₁₀O
MDL Number:MFCD00005367
:86.13 g/mol
Boiling Pt:78…82 °C
Melting Pt:–136 °C
Density:0.856 g/cm³ (25 °C)
Flash Pt:–11 °C
Storage Temperature:Ambient
EINECS:202-507-4
UN:2536
ADR:3,II
:GHS02
:GHS05
:GHS07