
4,6-Dichloro-2-methylpyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1780-26-3
:
Formula:C₅H₄Cl₂N₂
MDL Number:MFCD00090472
:163.01 g/mol
Melting Pt:41…45 °C
Flash Pt:97 °C
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05