Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl nicotinoylacetate
Methyl nicotinoylacetate
Methyl nicotinoylacetate
Irritant
Methyl nicotinoylacetate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl nicotinoylacetate
Supplier:
Apollo Scientific
About this item
CAS Number:
54950-20-8
:
Formula:
C₉H₉NO₃
MDL Number:
MFCD00216524
:
179.175 g/mol
Melting Pt:
34…42 °C
:
GHS07
Storage Temperature:
Ambient