Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dioxan-2-one
1,3-Dioxan-2-one
1,3-Dioxan-2-one
:
Apollo Scientific
1,3-Dioxan-2-one
:
Apollo Scientific
:
2453-03-4
:
:
C₄H₆O₃
:
MFCD00010440
:
135 °C/3.75mmHg
:
48 °C
:
>110 °C
:
102.09 g/mol
:
GHS07
:
Ambient