2,6-Dimethylbenzoic acid
Catalog # APOSOR41144-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:150.18 g/mol
Boiling Pt:160 °C/17mmHg
Melting Pt:114…117 °C
EINECS:211-177-0
:GHS07
Storage Temperature:Ambient
CAS Number:632-46-2
Formula:C₉H₁₀O₂
MDL Number:MFCD00002483