Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dihydroxybenzaldehyde

3,5-Dihydroxybenzaldehyde

Irritant
Supplier:   Apollo Scientific
3,5-Dihydroxybenzaldehyde
Supplier:  Apollo Scientific
Certificates
3,5-Dihydroxybenzaldehyde

2 Options Available Below

Product Details & Documents
  • CAS Number:26153-38-8
  • Formula:C₇H₆O₃
  • MDL Number:MFCD00016611
  • Molecular Weight:138.12 g/mol
  • Melting Pt:153…161 °C
  • Storage Temperature:Ambient
  • EINECS:247-479-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 43,50 €
    43,50 €
    1 * 1 g
       43,50 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 116,00 €
    116,00 €
    1 * 5 g
       116,00 €