
2-Oxa-7-azaspiro[3.5]nonane hemioxalate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1429056-28-9
:MFCD22422281
Formula:C₁₆H₂₈N₂O₆
MDL Number:MFCD11035888
:344.4 g/mol
Boiling Pt:208.4 °C
Density:1.048 g/cm³ (25 °C)
:GHS07
Storage Temperature:Refrigerator