Order Entry
Export
ContactUsLinkComponent
3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propionaldehyde 95%
Irritant
Environment
3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propionaldehyde 95%
Catalog # APOSOR40638-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Environment
3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propionaldehyde 95%
Catalog # APOSOR40638-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :203.2 g/mol
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:2436-29-5
Formula:C₁₁H₉NO₃
MDL Number:MFCD01704466