Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Dichloropyridine-2-carboxamide
3,6-Dichloropyridine-2-carboxamide
3,6-Dichloropyridine-2-carboxamide
3,6-Dichloropyridine-2-carboxamide
APOSOR40361-1G
:
Apollo Scientific
:
3,6-Dichloropyridine-2-carboxamide
APOSOR40361-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
191.02 g/mol
:
178…180 °C
:
GHS07
:
Ambient
:
1532-25-8
:
C₆H₄Cl₂N₂O
:
MFCD00186645