Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloro-4-hydroxypyridine
2,6-Dichloro-4-hydroxypyridine
2,6-Dichloro-4-hydroxypyridine
Irritant
2,6-Dichloro-4-hydroxypyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,6-Dichloro-4-hydroxypyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
17228-74-9
:
Formula:
C₅H₃Cl₂NO
MDL Number:
MFCD00955619
Melting Pt:
201…202 °C
:
163.9894 g/mol
:
GHS07
Storage Temperature:
Ambient