

2,4-Dichloro-5-iodopyrimidine
Catalog # APOSOR40189-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:274.87 g/mol
Melting Pt:65…69 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS06
:GHS05
CAS Number:13544-44-0
Formula:C₄HCl₂IN₂
MDL Number:MFCD01898087