Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloroimidazo[1,2-a]pyrazine
6-Chloroimidazo[1,2-a]pyrazine
6-Chloroimidazo[1,2-a]pyrazine
Irritant
6-Chloroimidazo[1,2-a]pyrazine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-Chloroimidazo[1,2-a]pyrazine
Supplier:
Apollo Scientific
About this item
CAS Number:
76537-23-0
:
Formula:
C₆H₄ClN₃
MDL Number:
MFCD09909644
:
153.5700 g/mol
:
GHS07
Storage Temperature:
Ambient