Order Entry
Export
ContactUsLinkComponent
2-(Benzyloxy)-4-bromoanisole
2-(Benzyloxy)-4-bromoanisole
Catalog # APOSOR400659-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(Benzyloxy)-4-bromoanisole
Catalog # APOSOR400659-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :293.1558 g/mol
Storage Temperature:Ambient
CAS Number:78504-28-6
Formula:C₁₄H₁₃BrO₂
MDL Number:MFCD09836381