1-Octanol 98%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: n-Octanol
- CAS Number:111-87-5
- Formula:H₃C(CH₂)₆CH₂OH
- MDL Number:MFCD00002988
- Molecular Weight:130.23 g/mol
- Boiling Pt:193…195 °C
- Melting Pt:–16 °C
- Density:0.825 g/cm³ (25 °C)
- Flash Pt:81 °C
- Storage Temperature:Ambient
- EINECS:203-917-6
4 Options Available Below
Product Details & Documents
Synonyms: n-Octanol
- CAS Number:111-87-5
- Formula:H₃C(CH₂)₆CH₂OH
- MDL Number:MFCD00002988
- Molecular Weight:130.23 g/mol
- Boiling Pt:193…195 °C
- Melting Pt:–16 °C
- Density:0.825 g/cm³ (25 °C)
- Flash Pt:81 °C
- Storage Temperature:Ambient
- EINECS:203-917-6
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



