Order Entry
Export
ContactUsLinkComponent
4-Bromo-5-methylbenzene-1,2-diamine
4-Bromo-5-methylbenzene-1,2-diamine
Catalog # APOSOR400118-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-5-methylbenzene-1,2-diamine
Catalog # APOSOR400118-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :201.07 g/mol
Storage Temperature:Ambient
CAS Number:102169-44-8
Formula:C₇H₉BrN₂
MDL Number:MFCD16987757