3-Acetylbenzophenone
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1-(3-Benzoylphenyl)ethan-1-one, 1-(3-Benzoylphenyl)ethanone
- CAS Number:66067-44-5
- Formula:C₁₅H₁₂O₂
- MDL Number:MFCD01662280
- Molecular Weight:224.2546 g/mol
- Boiling Pt:80…85 °C/0.1mmHg
- Density:1.22 g/cm³ (25 °C)
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
Synonyms: 1-(3-Benzoylphenyl)ethan-1-one, 1-(3-Benzoylphenyl)ethanone
- CAS Number:66067-44-5
- Formula:C₁₅H₁₂O₂
- MDL Number:MFCD01662280
- Molecular Weight:224.2546 g/mol
- Boiling Pt:80…85 °C/0.1mmHg
- Density:1.22 g/cm³ (25 °C)
- Storage Temperature:Ambient



