Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(3,4-Dimethoxyphenyl)-1H-pyrazole
3-(3,4-Dimethoxyphenyl)-1H-pyrazole
3-(3,4-Dimethoxyphenyl)-1H-pyrazole
Irritant
3-(3,4-Dimethoxyphenyl)-1H-pyrazole
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-(3,4-Dimethoxyphenyl)-1H-pyrazole
Supplier:
Apollo Scientific
About this item
CAS Number:
154257-70-2
:
Formula:
C₁₁H₁₂N₂O₂
MDL Number:
MFCD02091527
:
204.23 g/mol
:
GHS07
Storage Temperature:
Ambient