
3-[(4,6-Dimethoxypyrimidin-2-yl)oxy]benzoic acid
Catalog # APOSOR3995-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:276.25 g/mol
Melting Pt:206…208 °C
:GHS07
Storage Temperature:Ambient
CAS Number:387350-58-5
Formula:C₁₃H₁₂N₂O₅
MDL Number:MFCD00203067