
2,4-Dichloro-5-nitropyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:49845-33-2
:
Formula:C₄HCl₂N₃O₂
MDL Number:MFCD00127867
:193.98 g/mol
Boiling Pt:0.9 °C/0.5mmHg
Melting Pt:28…32 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Freezer