Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Di-tert-butylhydroquinone
2,5-Di-tert-butylhydroquinone
2,5-Di-tert-butylhydroquinone
Irritant
2,5-Di-tert-butylhydroquinone
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,5-Di-tert-butylhydroquinone
Supplier:
Apollo Scientific
About this item
CAS Number:
88-58-4
:
Formula:
C₁₄H₂₂O₂
MDL Number:
MFCD00008825
:
222.33 g/mol
Melting Pt:
217…219 °C
:
GHS07
Storage Temperature:
Ambient