2,7-Dimethylnaphthalene
Catalog # APOSOR3785-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:262…264 °C
Melting Pt:96…98 °C
:156.23 g/mol
Storage Temperature:Ambient
CAS Number:582-16-1
Formula:C₁₂H₁₂
MDL Number:MFCD00004121