0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
3,4-Dimethylphenylhydrazinium chloride
3,4-Dimethylphenylhydrazinium chloride 98%
3,4-Dimethylphenylhydrazinium chloride 98%
:
Apollo Scientific
3,4-Dimethylphenylhydrazinium chloride 98%
:
Apollo Scientific
:
60481-51-8
:
MFCD00052270
:
C₈H₁₃ClN₂
:
196 °C (dec.)
:
172.66 g/mol
:
GHS07
:
Refrigerator