Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6,8-Dichloroimidazo[1,2-a]pyridine
6,8-Dichloroimidazo[1,2-a]pyridine
6,8-Dichloroimidazo[1,2-a]pyridine
Corrosive
Irritant
6,8-Dichloroimidazo[1,2-a]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
6,8-Dichloroimidazo[1,2-a]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
858516-69-5
:
Formula:
C₇H₄Cl₂N₂
MDL Number:
MFCD06761692
:
187.03 g/mol
Melting Pt:
152…154 °C
:
GHS05
:
GHS07
Storage Temperature:
Refrigerator