
1,3-Dimethyl-1H-pyrazole-5-boronic acid 95+%
Catalog # APOSOR361166-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:139.9500 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:847818-68-2
Formula:C₅H₉BN₂O₂
MDL Number:MFCD12407383