3,5-Bis(methoxycarbonyl)phenylboronic acid 97%
Catalog # APOSOR360492-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:238.00 g/mol
Storage Temperature:Ambient
CAS Number:177735-55-6
Formula:C₁₀H₁₁BO₆
MDL Number:MFCD11053854