Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2'-Bibenzo[d][1,3,2]dioxaborole
2,2'-Bibenzo[d][1,3,2]dioxaborole
2,2'-Bibenzo[d][1,3,2]dioxaborole
:
Apollo Scientific
2,2'-Bibenzo[d][1,3,2]dioxaborole
:
Apollo Scientific
:
13826-27-2
:
:
C₁₂H₈B₂O₄
:
MFCD09842343
:
237.8115 g/mol
:
189…196 °C
:
GHS07
:
Ambient