Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dimethoxyphenylboronic acid pinacol ester
3,4-Dimethoxyphenylboronic acid pinacol ester 97%
3,4-Dimethoxyphenylboronic acid pinacol ester 97%
APOSOR360016-5G
:
Apollo Scientific
:
3,4-Dimethoxyphenylboronic acid pinacol ester 97%
APOSOR360016-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
264.13 g/mol
:
GHS07
:
Ambient
:
365564-10-9
:
C₁₄H₂₁BO₄
:
MFCD05863909