
3,4-Dimethoxybenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23095-31-0
:
Formula:C₈H₉ClO₄S
MDL Number:MFCD00051769
:236.68 g/mol
Melting Pt:67…70 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS05