Order Entry
Export
Orders LinkContactUsLinkComponent
2-(p-Tolyl)pyridine

2-(p-Tolyl)pyridine

Supplier:   Apollo Scientific
2-(p-Tolyl)pyridine
Supplier:  Apollo Scientific
Certificates
2-(p-Tolyl)pyridine

5 Options Available Below

Product Details & Documents
  • CAS Number:4467-06-5
  • Formula:C₁₂H₁₁N
  • MDL Number:MFCD00006281
  • Molecular Weight:169.22 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 47,80 €
    47,80 €
    1 * 25 g
       47,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 186,00 €
    186,00 €
    1 * 100 g
       186,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 921,00 €
    921,00 €
    1 * 500 g
       921,00 €

    Specifications:
    New Product
    New Product