trans-1,2-Cyclohexanedicarboxylic acid
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: trans-Cyclohexane-1,2-dicarboxylic acid
- CAS Number:2305-32-0
- Formula:C₈H₁₂O₄
- MDL Number:MFCD00001464
- Molecular Weight:172.18 g/mol
- Storage Temperature:Ambient
6 Options Available Below
Product Details & Documents
Synonyms: trans-Cyclohexane-1,2-dicarboxylic acid
- CAS Number:2305-32-0
- Formula:C₈H₁₂O₄
- MDL Number:MFCD00001464
- Molecular Weight:172.18 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



