Order Entry
Export
ContactUsLinkComponent
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 95%
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 95%
Catalog # APOSOR350342-25G
Supplier:  Apollo Scientific
CAS Number:  
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 95%
Catalog # APOSOR350342-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :213.28 g/mol
Storage Temperature:Ambient
CAS Number:23364-44-5
Formula:C₁₄H₁₅NO
MDL Number:MFCD00074959