Order Entry
Export
ContactUsLinkComponent
(4R,6S)-6-Methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol
(4R,6S)-6-Methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol
Catalog # APOSOR350339-5G
Supplier:  Apollo Scientific
CAS Number:  
(4R,6S)-6-Methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol
Catalog # APOSOR350339-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :218.29 g/mol
Storage Temperature:Ambient
CAS Number:147128-77-6
Formula:C₈H₁₀O₃S₂
MDL Number:MFCD11035925