Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Diaminopyrazine
2,3-Diaminopyrazine 97%
2,3-Diaminopyrazine 97%
2,3-Diaminopyrazine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2,3-Diaminopyrazine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
13134-31-1
:
Formula:
C₄H₆N₄
MDL Number:
MFCD07778385
:
110.1172 g/mol
Storage Temperature:
Refrigerator