4,4-Pentamethylene-2-pyrrolidone
Catalog # APOSOR350325-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:153.23 g/mol
Melting Pt:84…89 °C
Storage Temperature:Ambient
CAS Number:64744-50-9
Formula:C₉H₁₅NO
MDL Number:MFCD00177938