Pyridine-2,3-dicarboxylic anhydride
Catalog # APOSOR350318-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:149.11 g/mol
Storage Temperature:Ambient
CAS Number:699-98-9
Formula:C₇H₃NO₃
MDL Number:MFCD00005915