1-Methyl-2-piperidinemethanol
Catalog # APOSOR350237-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:129.2 g/mol
Storage Temperature:Ambient
CAS Number:20845-34-5
Formula:C₇H₁₅NO
MDL Number:MFCD00006494