Methyl-3-piperidinecarboxylate
Catalog # APOSOR350220-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:143.1836 g/mol
Storage Temperature:Ambient
CAS Number:50585-89-2
Formula:C₇H₁₃NO₂
MDL Number:MFCD00797850