2,6-Dichlorocinnamic acid
Catalog # APOSOR350156-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:217.05 g/mol
Storage Temperature:Ambient
CAS Number:5345-89-1
Formula:C₉H₆Cl₂O₂
MDL Number:MFCD00004374