Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Dimethylpyrazole

1,4-Dimethylpyrazole

Supplier:   Apollo Scientific
1,4-Dimethylpyrazole
Supplier:  Apollo Scientific
Certificates
1,4-Dimethylpyrazole

5 Options Available Below

Product Details & Documents
  • CAS Number:1072-68-0
  • Formula:C₅H₈N₂
  • MDL Number:MFCD01075158
  • Molecular Weight:96.1300 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 30,40 €
    30,40 €
    1 * 25 g
       30,40 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 89,80 €
    89,80 €
    1 * 100 g
       89,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 385,00 €
    385,00 €
    1 * 500 g
       385,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 890,00 €
    1 890,00 €
    1 * 2,5 kg
       1 890,00 €

    Specifications:
    New Product
    New Product