Order Entry
Export
ContactUsLinkComponent
3,3-Diphenylpropionic acid
3,3-Diphenylpropionic acid
Catalog # APOSOR350117-100G
Supplier:  Apollo Scientific
CAS Number:  
3,3-Diphenylpropionic acid
Catalog # APOSOR350117-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :226.27 g/mol
Storage Temperature:Ambient
CAS Number:606-83-7
Formula:C₁₅H₁₄O₂
MDL Number:MFCD00002717