
2,2-Dimethylsuccinic anhydride
Catalog # APOSOR350114-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:128.1259 g/mol
Boiling Pt:219…220 °C
Melting Pt:26…30 °C
Flash Pt:108 °C
:GHS07
Storage Temperature:Ambient
CAS Number:17347-61-4
Formula:C₆H₈O₃
MDL Number:MFCD00022608