Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(tert-Butoxycarbonyl)-D-phenylalanine
N-(tert-Butoxycarbonyl)-D-phenylalanine
N-(tert-Butoxycarbonyl)-D-phenylalanine
Irritant
N-(tert-Butoxycarbonyl)-D-phenylalanine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
N-(tert-Butoxycarbonyl)-D-phenylalanine
Supplier:
Apollo Scientific
About this item
CAS Number:
18942-49-9
:
Formula:
C₁₄H₁₉NO₄
MDL Number:
MFCD00063149
:
265.31 g/mol
Melting Pt:
78…87 °C
:
GHS07
Storage Temperature:
Refrigerator