Order Entry
Export
ContactUsLinkComponent
2,5-Dichloro-3-methylpyridine 97%
2,5-Dichloro-3-methylpyridine 97%
Catalog # APOSOR3474-5G
Supplier:  Apollo Scientific
CAS Number:  
2,5-Dichloro-3-methylpyridine 97%
Catalog # APOSOR3474-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :162.0166 g/mol
Storage Temperature:Ambient
CAS Number:59782-88-6
Formula:C₆H₅Cl₂N
MDL Number:MFCD03840754