Order Entry
Export
ContactUsLinkComponent
3-Bromo[1,2,4]triazolo[4,3-a]pyridine 95+%
3-Bromo[1,2,4]triazolo[4,3-a]pyridine 95+%
Catalog # APOSOR346605-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo[1,2,4]triazolo[4,3-a]pyridine 95+%
Catalog # APOSOR346605-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.0200 g/mol
Storage Temperature:Ambient
CAS Number:4922-68-3
Formula:C6H4BrN3
MDL Number:MFCD11111699