Order Entry
Export
Orders LinkContactUsLinkComponent
3-Bromopyrazolo[1,5-a]pyridine 95+%

3-Bromopyrazolo[1,5-a]pyridine 95+%

Bottle, 1 g
Catalog # APOSOR346595-1G
Supplier:   Apollo Scientific
3-Bromopyrazolo[1,5-a]pyridine 95+%
Catalog # APOSOR346595-1G
Supplier:  Apollo Scientific
CAS Number:  5910-12-3
3-Bromopyrazolo[1,5-a]pyridine 95+%

Bottle, 1 g

CAS Number:  5910-12-3
Price
304,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:5910-12-3
  • Formula:C₇H₅BrN₂
  • MDL Number:MFCD06661764
  • Molecular Weight:197.03 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications