Order Entry
Export
ContactUsLinkComponent
Imidazo[2,1-b]thiazol-6-ylmethanol
Imidazo[2,1-b]thiazol-6-ylmethanol
Catalog # APOSOR346325-1G
Supplier:  Apollo Scientific
CAS Number:  
Imidazo[2,1-b]thiazol-6-ylmethanol
Catalog # APOSOR346325-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :154.190 g/mol
Storage Temperature:Ambient
CAS Number:349480-74-6
Formula:C₆H₆N₂OS
MDL Number:MFCD07368518