Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Phenyl-4,5-dihydropyridazin-3(2H)-one
6-Phenyl-4,5-dihydropyridazin-3(2H)-one
6-Phenyl-4,5-dihydropyridazin-3(2H)-one
6-Phenyl-4,5-dihydropyridazin-3(2H)-one
Supplier:
Apollo Scientific
Certificates
Expand image
6-Phenyl-4,5-dihydropyridazin-3(2H)-one
Supplier:
Apollo Scientific
About this item
CAS Number:
1011-46-7
:
Formula:
C₁₀H₁₀N₂O
MDL Number:
MFCD00099597
:
174.2 g/mol
Storage Temperature:
Ambient