Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Thien-3-ylcyclohexane-1,3-dione
5-Thien-3-ylcyclohexane-1,3-dione
5-Thien-3-ylcyclohexane-1,3-dione
APOSOR346279-1G
:
Apollo Scientific
:
5-Thien-3-ylcyclohexane-1,3-dione
APOSOR346279-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
194.25 g/mol
:
Ambient
:
239132-57-1
:
C₁₀H₁₀O₂S
:
MFCD12827893