2,4-Dibromopentanedioic acid
Catalog # APOSOR346158-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:289.92 g/mol
Storage Temperature:Ambient
CAS Number:3479-80-9
Formula:C₅H₆Br₂O₄
MDL Number:MFCD11845705