N'-[1-Amino-2-(2-chlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1053655-91-6
:
Formula:C₁₃H₁₈ClN₃O₃
MDL Number:MFCD10568189
:299.76 g/mol
Storage Temperature:Ambient